31801.18
31801.30
31801.A1
31801.18EA
27.3
EUR
InStock
31801.18
31801.30
31801.A1
Tris(hydroxymethyl)aminomethan (TRIS, Trometamol) 99,8-100,1% (Trockenmasse) ACS
Tris(hydroxymethyl)aminomethan (TRIS, Trometamol)
Purity: 99.8-100.1%
CAS No.: 77-86-1
Molecular Formula: (HOCH2)3CNH2
Formula Weight: 121.14
Physical Form: Crystalline
Density: 1.353
Melting Point: 171-172°
Boiling Point: 219-220°/10mm
Beilstein Reference: 741883
Merck Reference: 14,9772
MDL No.: MFCD00004679