Walk-in-chambers for Stability Testing and Storage, WIC
Lieferant: Binder
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
9800-0011 9800-0012 9800-0013
390-1702EA 74900 EUR InStock
390-1702 390-1703 390-1704
Walk-in-chambers for Stability Testing and Storage, WIC
Prüfkammern Klimakammern
The WIC series walk-in-chambers offer a wide range of features and benefits to perfectly meet requirements in stability testing. Perfect test conditions are maintained over a wide range of temperatures from 10 to 50 °C and relative humidity of 20 to 90% RH.
  • Units have relative humidity accuracy of ±2,5% and temperature accuracy of ±1,5 °C
  • Duct work for proper distribution of air
  • Highly insulated room panels with U value of only 0,21 W/min⁻² K
  • LED lighting with motion detector
  • High quality room elements, made in Germany
  • Interior completely stainless steel
  • Exterior service accessibility
Room Elements: Galvanized sheet steel, polyester coated, white, similar to RAL 9010, interior surfaces in stainless steel, 100 mm PU foam U value: 0,21 W/min⁻² K. Floor overlay is chrome-nickel steel, pattern-rolled 5WL. Surface load: 50000 N/m⁻².
The Binder walk-in chambers offer a wide range of features and benefits to perfectly meet requirements in stability testing. Choice of 6, 9 or 12 m² internal dimensions and 2,3 m height are an excellent option for stability testing in pharma, food and beverage, cosmetics, material testing and many other fields of application.
Perfect test conditions are maintained in a wide range of temperature from 10 to 50 °C and relative humidity of 20 to 90% RH. In Pharma, the Walk-in chambers cover long-term and accelerated stability test conditions as specified in the ICH Q1A (R2) for new drug substances and drug products.
Power frequency: 50 Hz
Unit fuze: 1 A
Phase (nominal voltage): 1~
Electrical: 200 to 230 V, 50/60 Hz, 1~
Humidity fluctuation: ±2,5% at 25 °C and 60% RH
Zertifizierungen: IP protection class according to DIN EN 60529: IP 20.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge