Crucibles, round bottom, zirconium oxide
Lieferant: Alfa Aesar
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
41297.KF 41300.KF
41297.KFEA 236 EUR InStock
41297.KF 41300.KF
Crucibles, round bottom, zirconium oxide
Tiegel
These crucibles are manufactured from yttria stabilised zirconium oxide. They display superior characteristics both at elevated temperature and at high frequencies, and are resistant to corrosive liquids and gases. Oxide ceramics are also inert to oxidation and are not subject to radiation damage.
  • Increased working temperature characteristics
  • Offer useful alternatives to aluminium
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge