A17220.30
A17220.0B
A17220.30EA
29.6
EUR
InStock
A17220.30
A17220.0B
3,3'-Thiodipropionsäure ≥98%
3,3'-Thiodipropionsäure
Purity: 98%
CAS No.: 111-17-1
Molecular Formula: C6H10O4S
Formula Weight: 178.21
Melting Point: 127-134°
Beilstein Reference: 1210299
Merck Reference: 14,9332
MDL No.: MFCD00002781