Recombinat Protein,Human BMPR-IA ALK-3 Protein, Fc Tag,Human BMPR-IA, Fc Tag (AL3-H5259) is expressed from human 293 cells (HEK293) . It contains AA Gln 24 - Arg 152 (Accession CAA80257).This protein carries a human IgG1 Fc tag at the C-terminus. Th 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human BMPR-IA ALK-3 Protein, Fc Tag,Human BMPR-IA, Fc Tag (AL3-H5259) is expressed from human 293 cells (HEK293) . It contains AA Gln 24 - Arg 152 (Accession CAA80257).This protein carries a human IgG1 Fc tag at the C-terminus. Th 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human BMPR-IA ALK-3 Protein, Fc Tag,Human BMPR-IA, Fc Tag (AL3-H5259) is expressed from human 293 cells (HEK293) . It contains AA Gln 24 - Arg 152 (Accession CAA80257).This protein carries a human IgG1 Fc tag at the C-terminus. Th 1 * 1 mg ACRBAL3-H5259-1MG 1 * 1 mg
Hersteller-Artikelnummer:: AL3-H5259-1MG
Lokale Artikelnummer:: ACRBAL3-H5259-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder