Recombinat Protein,Human BTN3A3 BTF3 Protein, His Tag,Human BTN3A3, His Tag (BT3-H52H0) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Trp 248 (Accession AAH15815).This protein carries a polyhistidine tag at the C-terminus. The 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human BTN3A3 BTF3 Protein, His Tag,Human BTN3A3, His Tag (BT3-H52H0) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Trp 248 (Accession AAH15815).This protein carries a polyhistidine tag at the C-terminus. The 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human BTN3A3 BTF3 Protein, His Tag,Human BTN3A3, His Tag (BT3-H52H0) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Trp 248 (Accession AAH15815).This protein carries a polyhistidine tag at the C-terminus. The 1 * 1 mg ACRBBT3-H52H0-1MG 1 * 1 mg
Hersteller-Artikelnummer:: BT3-H52H0-1MG
Lokale Artikelnummer:: ACRBBT3-H52H0-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder