Recombinat Protein,Mouse BTLA Protein, Fc Tag,Mouse BTLA, Fc Tag (BTA-M5253) is expressed from human 293 cells (HEK293). It contains AA Glu 30 - Gly 176 (Accession Q32MV9).This protein carries a human IgG1 Fc tag at the C-terminus. The protein has o 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Mouse BTLA Protein, Fc Tag,Mouse BTLA, Fc Tag (BTA-M5253) is expressed from human 293 cells (HEK293). It contains AA Glu 30 - Gly 176 (Accession Q32MV9).This protein carries a human IgG1 Fc tag at the C-terminus. The protein has o 1 * 100 G
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Mouse BTLA Protein, Fc Tag,Mouse BTLA, Fc Tag (BTA-M5253) is expressed from human 293 cells (HEK293). It contains AA Glu 30 - Gly 176 (Accession Q32MV9).This protein carries a human IgG1 Fc tag at the C-terminus. The protein has o 1 * 100 µG ACRBBTA-M5253100UG 1 * 100 µG
Hersteller-Artikelnummer:: BTA-M5253100UG
Lokale Artikelnummer:: ACRBBTA-M5253100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder