Recombinat Protein,Human E-Selectin CD62E Protein, His Tag,Human E-Selectin, His Tag (CDE-H5222) is expressed from human 293 cells (HEK293). It contains AA Trp 22 - Pro 556 (Accession NP000441.2).This protein carries a polyhistidine tag at the C-ter 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human E-Selectin CD62E Protein, His Tag,Human E-Selectin, His Tag (CDE-H5222) is expressed from human 293 cells (HEK293). It contains AA Trp 22 - Pro 556 (Accession NP000441.2).This protein carries a polyhistidine tag at the C-ter 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human E-Selectin CD62E Protein, His Tag,Human E-Selectin, His Tag (CDE-H5222) is expressed from human 293 cells (HEK293). It contains AA Trp 22 - Pro 556 (Accession NP000441.2).This protein carries a polyhistidine tag at the C-ter 1 * 1 mg ACRBCDE-H5222-1MG 1 * 1 mg
Hersteller-Artikelnummer:: CDE-H5222-1MG
Lokale Artikelnummer:: ACRBCDE-H5222-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder