Human Ephrin-A4, Fc Tag (EA4-H5250) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Gly 171 (Accession AAI07484 ). 1 * 250 G
Lieferant: ACROBIOSYSTEMS
Human Ephrin-A4, Fc Tag (EA4-H5250) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Gly 171 (Accession AAI07484 ). 1 * 250 G
PRODUKT
Erweitert / Kompakt
Human Ephrin-A4, Fc Tag (EA4-H5250) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Gly 171 (Accession AAI07484 ). 1 * 250 µG ACRBEA4-H5250250UG 1 * 250 µG
Hersteller-Artikelnummer:: EA4-H5250250UG
Lokale Artikelnummer:: ACRBEA4-H5250250UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder