Human G-CSF R, Fc Tag (GCR-H5250) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Pro 621 (Accession NP000751.1 ). 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Human G-CSF R, Fc Tag (GCR-H5250) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Pro 621 (Accession NP000751.1 ). 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Human G-CSF R, Fc Tag (GCR-H5250) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Pro 621 (Accession NP000751.1 ). 1 * 1 mg ACRBGCR-H5250-1MG 1 * 1 mg
Hersteller-Artikelnummer:: GCR-H5250-1MG
Lokale Artikelnummer:: ACRBGCR-H5250-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder