Biotinylated Human IL-2 R alpha, His,Avitag (ILA-H82E6) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Cys 213 (Accession P01589-1). 1 * 25 G
Lieferant: ACROBIOSYSTEMS
Biotinylated Human IL-2 R alpha, His,Avitag (ILA-H82E6) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Cys 213 (Accession P01589-1). 1 * 25 G
PRODUKT
Erweitert / Kompakt
Biotinylated Human IL-2 R alpha, His,Avitag (ILA-H82E6) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Cys 213 (Accession P01589-1). 1 * 25 µG ACRBILA-H82E6-25UG 1 * 25 µG
Hersteller-Artikelnummer:: ILA-H82E6-25UG
Lokale Artikelnummer:: ACRBILA-H82E6-25UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder