Human IL-2 R beta, Fc Tag (SPR verified) (ILB-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 (Ac cession NP000869.1). 1 * 50 G
Lieferant: ACROBIOSYSTEMS
Human IL-2 R beta, Fc Tag (SPR verified) (ILB-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 (Ac cession NP000869.1). 1 * 50 G
PRODUKT
Erweitert / Kompakt
Human IL-2 R beta, Fc Tag (SPR verified) (ILB-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 (Ac cession NP000869.1). 1 * 50 µG ACRBILB-H5253-50UG 1 * 50 µG
Hersteller-Artikelnummer:: ILB-H5253-50UG
Lokale Artikelnummer:: ACRBILB-H5253-50UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder