B22778.14
B22778.22
B22778.36
B22778.14EA
34.3
EUR
InStock
B22778.14
B22778.22
B22778.36
α,4-Dichlor-α-phenyltoluol 97%
α,4-Dichlor-α-phenyltoluol
Purity: 98%
CAS No.: 134-83-8
Molecular Formula: C13H10Cl2
Formula Weight: 237.13
Flash Point: 194°(381°F)
Refractive Index: 1.6030
Boiling Point: 159-160°/2mm
Beilstein Reference: 2050236
UN No.: 3265
MDL No.: MFCD00000856