Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NCAM2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-11094R-A350
BOSSBS-11094R-A350EA 893 EUR InStock
BOSSBS-11094R-A350
Anti-NCAM2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
NCAM2 is an 837 amino acid protein encoded by the human gene NCAM2. NCAM2 contains five immunoglobulin-like domains, two Fibronectin type III domains, a transmembrane domain and a cytoplasmic domain. The gene is expressed most strongly in human adult and fetal brain. NCAM2 is a member of the neural cell adhesion molecule (NCAM) family. NCAMs are closely related cell surface glycoproteins involved in cell to cell interactions during growth and are thought to play an important role in embryogenesis and development. NCAM2 is a considered a good candidate for involvement in certain Down syndrome phenotypes because a slight overexpression of NCAMs increases many-fold the homotypic adhesion properties of cells. Stat5 regulates NCAM2 in vivo by binding to the NCAM2 intron in the NKL natural killer cell line; this binding is induced by cytokines that activate Stat5. Neither Stat1 nor Stat3 bind to this region, despite sharing a consensus binding sequence with Stat5.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: NCAM2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge