Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TROVE2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-7116R-FITC
BOSSBS-7116R-FITCEA 900 EUR InStock
BOSSBS-7116R-FITC
Anti-TROVE2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Ro autoantigens are of clinical significance because directed against them are found in most patients with primary Sjqgren syndrome, subacute cutaneous lupus erythematosus (SLE), neonatal lupus erythematosus, ANA-negative lupus erythematosus, and systemic lupus erythematosus-like disease secondary to homozygous C2 or C4 complement deficiency (1). Ro/SSA is a ribonucleoprotein that binds to auto in 35 to 50% of patients with SLE and in up to 97% of patients with Sjqgren syndrome (2). The Ro/SSA particle consists of a single immunoreactive protein noncovalently bound with one of four small RNA molecules (2). Most anti-Ro/SSA-positive sera detect not only the main protein, but also a smaller Ro/SSA protein (2). The genes which encode the smaller and larger proteins map to human chromosomes 11p15.5 and 1q31, respectively (3?). La/SSB is an autoimmune RNA-binding protein that plays a role in the transcription of RNA polymerase III was originally defined by its reactivity with auto from patients with Sjé°ƒren syndrome and SLE (6).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TROVE2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge