Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ARG1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-8585R-CY5
BOSSBS-8585R-CY5EA 922 EUR InStock
BOSSBS-8585R-CY5
Anti-ARG1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Arginase I which is expressed almost exclusively in the liver, catalyzes the conversion of arginine to ornithine and urea . The human arginase I gene, which maps to chromosome 6q23, encodes a 322 amino acid protein. Arginase I exists as a homotrimeric protein and contains a binuclear manganese cluster. Arginase II catalyzes the same reaction as arginase I, but differs in its tissue specificity and subcellular location. Specifically, arginase II localizes to the mitochondria. Arginase II is expressed in non-hepatic tissues, with the highest levels of expression in the kidneys, but, unlike arginase I, is not expressed in liver. The human arginase II gene, which maps to chromosome 14q24.1-q24.3, encodes a 354 amino acid protein. In addition, arginase II contains a putative amino-terminal mitochondrial localization sequence.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ARG1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge