Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CD300C Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-9203R-CY3
BOSSBS-9203R-CY3EA 1350 EUR InStock
BOSSBS-9203R-CY3
Anti-CD300C Rabbit Polyclonal Antibody (Cy3®)
Antikörper
CD300C , also known as CLM-6 (CMRF35-like molecule 6), is a 224 amino acid single-pass type I membrane protein that is present on the surface of neutrophils, monocytes and a select proportion of peripheral blood B and T lymphocytes. CD300C contains two potential N-gylcosylation sites, a potential O-glycosylated hinge-like region and an Ig-like V-type (immunoglobulin-like) domain that is very similar to the Fc receptor for polymeric IgA and IgM. The gene encoding CD300C is localized to a region on human chromosome 17 that harbors a susceptilibty locus for psoriasis, dermatitis and rheumatoid arthritis, suggesting a possible involvement of CD300C with these conditions.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD300C
Clonality: Polyclonal
Clone: 0
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge