Anti-Vimentin Chicken Polyclonal Antibody
Lieferant: Biosensis
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C-1409-50
BSENC-1409-50EA 312 EUR InStock
BSENC-1409-50
Anti-Vimentin Chicken Polyclonal Antibody
Antikörper
Vimentins are class-III intermediate filaments specific to mesenchymal tissue. Vimentin is an important cytoskeletal component responsible for maintaining cell integrity and has a probable role in the intracellular transport of proteins such as lipoproteins between the nucleus and plasma membrane. Immunohistochemical staining for Vimentin is characteristic of sarcomas. 
The specificity of this antibody has been confirmed by WB. This antibody detects ~50 kDa Vimentin protein in multiple species. The antibody also reacts excellently in immunocytochemistry and immunhistochemistry.
More Product Information
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge
Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.
Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..