Trimyristin 95%
Lieferant: ALFA AESAR
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
J62819.14
J62819.14EA 237 EUR InStock
J62819.14
Trimyristin 95%
Trimyristin
Purity: 95% CAS No.: 555-45-3 Molecular Formula: C45H86O6 Formula Weight: 723.16 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 56-57° Solubility: Insoluble in water. Soluble in chloroform, ether, ethanol or dichloromethane Beilstein Reference: 1718242 MDL No.: MFCD00036229
Formel: C₄₅H₈₆O₆
MW: 723.18 g/mol
Melting Pt: 56…57 °C
MDL Number: MFCD00036229
CAS-Nummer: 555-45-3
EINECS: 209-099-7
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge