Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Sulpho-NHS-LC-Biotin, EZ-Link™ No-Weigh™, Pierce™

Lieferant: Thermo Scientific

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


A39257
PIERA39257EA 236 EUR InStock
PIERA39257
Sulpho-NHS-LC-Biotin, EZ-Link™ No-Weigh™, Pierce™
Sulpho-NHS-LC-Biotin
Thermo Scientific EZ-Link Sulfo-NHS-LC-Biotin is an intermediate-length, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines.

  • Protein labeling—biotinylate antibodies to facilitate immobilization, purification or detection using streptavidin resins or probes
  • Cell surface labeling—biotinylates only surface proteins of whole cells because the negatively charged reagent does not permeate cell membranes
  • Amine-reactive—reacts with primary amines (-NH2), such as lysine side-chains, or the amino-termini of polypeptides
  • Soluble—charged sulfo-NHS group increases reagent water solubility compared to ordinary NHS-ester compounds
  • Irreversible—forms permanent amide bonds; spacer arm cannot be cleaved
  • Medium length—spacer arm (total length added to target) is 22.4 angstroms; provides excellent balance between adequate length (minimal steric hindrance for biotin binding) and modest mass

Sulfo-NHS-LC-Biotin is one of three very similar EZ-Link Reagents that are water-soluble, non-cleavable, and enable simple and efficient biotinylation of antibodies, proteins and any other primary amine-containing macromolecules in solution. Specific labeling of cell surface proteins is another common application for these uniquely water-soluble and membrane impermeable reagents. Differing only in their spacer arm lengths, the three Sulfo-NHS-ester reagents offer the possibility of optimizing labeling and detection experiments where steric hindrance of biotin binding is an important factor. Sulfo-NHS-LC-Biotin is offered in several package sizes and as complete protein labeling kits.

Thermo Scientific No-Weigh products are specialty reagents provided in a pre-aliquoted format. The pre-weighed packaging prevents the loss of reagent reactivity and contamination over time by eliminating the repetitive opening and closing of the vial. The format enables use of a fresh vial of reagent each time, eliminating the hassle of weighing small amounts of reagents and reducing concerns over reagent stability.

Achtung! Bitte beachten: For Research Use Only. Not for use in diagnostic procedures. Store desiccated at -20°C.


Formel: C₂₀H₂₉N₄NaO₉S₂
MW: 556.59 g/mol
Storage Temperature: Gefrierschrank
MDL Number: MFCD00078534
CAS-Nummer: 127062-22-0
Order Now
Spezifikation Testergebnisse
Product Type Sulfo-NHS-LC-Biotin
Quantity 10 x 1 mg
Labeling Method Chemical Labeling
Shipping Condition Qualified for Ambient or Wet Ice
Product Line EZ-Link™
Cell Permeability Cell-Impermeant
Chemical Reactivity Amine
Reactive Moiety NHS Ester
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or Dye Biotin
Spacer Mid-Length

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge