Thorium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Lieferant: PROMOCHEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
VHG-PTHN-100 VHG-PTHN-50 VHG-PTHN-500 VHG-TTHN-100 VHG-TTHN-500
PROOVHG-PTHN-100EA 58 EUR InStock
PROOVHG-PTHN-100 PROOVHG-PTHN-50 PROOVHG-PTHN-500 PROOVHG-TTHN-100 PROOVHG-TTHN-500
Thorium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Standards Kationischer Standards Thorium-Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Zertifizierungen: Traceable to NIST, SRM.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge