VHG-PTHN-100
VHG-PTHN-50
VHG-PTHN-500
VHG-TTHN-100
VHG-TTHN-500
PROOVHG-PTHN-100EA
58
EUR
InStock
PROOVHG-PTHN-100
PROOVHG-PTHN-50
PROOVHG-PTHN-500
PROOVHG-TTHN-100
PROOVHG-TTHN-500
Thorium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Standards
Kationischer Standards
Thorium-Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Zertifizierungen: Traceable to NIST, SRM.