Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

ortho-Phosphorsäure 85 - 87% ≥85,0-87,0%, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: AVANTOR PERFORMANCE MATERIALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

Veränderungsmanagement-Kategorie = N

0260-08EA 2410.66 EUR InStock
0260-08 0260-07 0260-15 0260-03 0260-01 0260-33 0260-06
ortho-Phosphorsäure 85 - 87% ≥85,0-87,0%, BAKER ANALYZED® ACS, J.T.Baker®
ortho-Phosphorsäure
The quality of BAKER ANALYZED™ ACS Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These acids are tested for common trace metals.


Formel: H₃PO₄
Melting Pt: 28 °C
Dichte: 1,69 g/cm³
Storage Temperature: Raumtemperatur
MDL Number: MFCD00011340
CAS-Nummer: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III
Order Now
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements 85.0 - 87.0%
For Laboratory,Research,or Manufacturing Use ≤0.002%
Assay (H₃PO₄) (by acidimetry) 85.0 - 87.0 %
Calcium (Ca) ≤ 0.002 %
Color (APHA) ≤ 10
Insoluble Matter ≤ 0.001 %
ACS - Magnesium (Mg) ≤ 0.002 %
Sulfate (SO₄) ≤ 12 ppm
Volatile Acids (as CH₃COOH) ≤ 0.001 %
Reducing Substances Passes Test
Chloride (Cl) ≤ 3 ppm
Nitrate (NO₃) ≤ 5 ppm
Trace Impurities - Antimony (Sb) ≤ 20.000 ppm
Trace Impurities - Arsenic (As) ≤ 0.500 ppm
Trace Impurities - Iron (Fe) ≤ 10 ppm
Heavy Metals (as Pb) ≤ 8 ppm
Trace Impurities - Manganese (Mn) ≤ 0.500 ppm
Trace Impurities - Potassium (K) ≤ 40.000 ppm
Trace Impurities - Sodium (Na) ≤ 200.000 ppm
Exceeds A.C.S. Specifications
Meets Reagent Specifications for testing USP/NF monographs

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge