10459.14
10459.22
10459.36
10459.14EA
45.1
EUR
InStock
10459.14
10459.22
10459.36
Aluminiumoxid, γ-α-Phase ≥99,98% (Metall-Basis)
Aluminiumoxid, γ-α-Phase
Purity: 99.98%
CAS No.: 1344-28-1
Molecular Formula: Al2O3
Formula Weight: 101.96
Physical Form: 0.01-0.02 Micron Powder, S.A. 100m2/g
Density: 3.965
Melting Point: 2045°
Boiling Point: 2980°
Merck Reference: 14,356
MDL No.: MFCD00003424