10664.14
10664.22
10664.36
10664.A3
10664.14EA
128
EUR
InStock
10664.14
10664.22
10664.36
10664.A3
Cadmium(II)oxid ≥99,998% (Metall-Basis), Puratronic®
Cadmium(II)oxid
Purity: 99.998%
CAS No.: 1306-19-0
Molecular Formula: CdO
Formula Weight: 128.40
Physical Form: Powder
Density: 8.15
Appearance: Red-brown
Odour: Odourless
Melting Point: 1430°
Merck Reference: 14,1623
UN No.: 2570
MDL No.: MFCD00010921