Safety faceshields, Shape™
Lieferant: JSP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
5SHA20C 5SHA2GEN20C
111-2594EA 29.3 EUR InStock
111-2594 111-2595
Safety faceshields, Shape™
Gesichtsschutz
A compact, comfortable, and lightweight faceshield with superior optical quality.
  • Perfect optical quality
  • 2,2 mm polycarbonate visor
  • Wide field of vision
  • Adjustable wheel ratchet headband
  • Compatible with prescription glasses
The Shape™ Safety Faceshield features a moulded polycarbonate visor that provides superior optical quality with an exceptional field of vision.
Zertifizierungen: EN 166; EN 170.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge