Gas goggle, non-vented, EVO®
Lieferant: JSP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AGM030-623-000
111-2596EA 24.9 EUR InStock
111-2596
Gas goggle, non-vented, EVO®
Vollsichtbrillen
Single lens goggle for everyday protection.
  • PVC frame
  • Elastic headband
  • Anti-mist
  • Molten metal protection
  • Non vented design
  • High level of protection
Standard single lens goggle for everyday protection, engineered with anti-mist and scratch resistant single lens with UV protection. Low profile design is lightweight with soft EVA foam housing for exceptional wearer comfort. Goggles offer dust, liquid splash, molten metal, and extreme temperature protection. Compatible with prescription lenses by using RX Inserts for EVO® Goggles.
Zertifizierungen: EN 166; EN 170.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge