11332.09
11332.22
11332.36
11332.09EA
25.3
EUR
InStock
11332.09
11332.22
11332.36
Ammoniumcer(IV)nitrat ≥99,5% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Ammoniumcer(IV)nitrat
Purity: 99.5%
CAS No.: 16774-21-3
Molecular Formula: H8CeN8O18
Formula Weight: 548.23
Physical Form: Crystalline
Appearance: Orange
Odour: Pungent
Merck Reference: 14,1992
UN No.: 1477
MDL No.: MFCD00151121