Porous polymer adsorbent, Tenax®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
11982
11982.EA 453 EUR InStock
11982.
Porous polymer adsorbent, Tenax®
Chromatographie-Sorbentien GC-Sorbentien
These porous polymers, based on 2,6-diphenyl-p-phenylene oxide, are widely used as an adsorbent in both air collection and purge trap applications.
The unique structure provides alternate and desirable adsorption/desorption characteristics compared to other porous polymers.
Surface area: ~35 m²/g
Density: ~0,25 g/ml (free fall density)
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge