12630.AD
12630.AK
12630.AU
12630.ADEA
116
EUR
InStock
12630.AD
12630.AK
12630.AU
Ruthenium(III) nitrosylnitrate -Lösung (1,5% Ru) (w/v)
Ruthenium(III) nitrosylnitrate
CAS No.: 34513-98-9
Molecular Formula: N4O10Ru
Formula Weight: 317.09
Physical Form: Liquid
Density: 1.07
Appearance: Red-brown
Odour: Acidic
UN No.: UN3093
MDL No.: MFCD00016313