Portable gas detectors, GasAlert Micro 5
Lieferant: BW TECHNOLOGIES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
M5IRXWBYRDDYE0
151-0248EA 0 EUR InStock
151-0248
Portable gas detectors, GasAlert Micro 5
Gas-Spürsysteme
Continuously monitoring up to five gases, the GasAlert Micro 5 was designed and engineered for an extensive range of applications.
  • LCD shows simultaneous gas concentrations
  • Integral motorized pump option for remote sampling
  • Two power options: Alkaline batteries or rechargeable pack
  • Multi-language support in English, French, German, Spanish, and Portuguese
Zertifizierungen: EMI/RFI: EMC 89/336/EEC; IP 65/66; ATEX: X g II 1 G Ga Ex ia IIC T4* X g II 2 G (IR model only); Ex d ia IIC T4* - IECEx: Ga Ex ia IIC T4* Ex d ia IIC T4* (IR model only)
* Temperature codes may vary as a function of the batteries installed
Lieferumfang: Supplied with specified sensor(s), stainless steel alligator clip and concussion-proof yellow housing, rechargeable battery pack or alkaline pack with three AA batteries, cradle charger and wall outlet charging adaptor with rechargeable battery option, screwdriver, calibration/test adaptor and hose, manual and multi-language CD manual.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge