16245.06
16245.14
16245.06EA
37.7
EUR
InStock
16245.06
16245.14
Thymolphthalein ACS
Thymolphthalein
CAS No.: 125-20-2
Molecular Formula: C28H30O4
Formula Weight: 430.55
Physical Form: Crystalline
Melting Point: 251-253°
Solubility: Insoluble in water. Soluble in alcohol, acetone, dilute alkalies (blue Colour), H2SO4 (red Colour)
Beilstein Reference: 359413
Merck Reference: 14,9401
MDL No.: MFCD00005909
pH indicator; Colourless below pH 9.3, blue above pH 10.5