18597-100ML
18597-500ML
18597-100MLEA
118
EUR
InStock
18597-100ML
18597-500ML
Triethylammonium bicarbonate buffer, for HPLC, LiChropur™, Supelco®
Puffer
Chromatographie-Puffer
1M Buffer, also for use in ion-exchange chromatography and electrophoresis.