1895-1AMP
1895-1AMPEA
126
EUR
InStock
1895-1AMP
Supelco® Organic Reference Standard, F.A.M.E. Mix Glc-40
Standards
Organische Standards
Zertifizierungen: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.
Verpackung: 100 mg ampule contains approximately equal amounts by weight of: Palmitic acid methyl ester (C16:0) Stearic acid methyl ester (C18:0) Arachidic acid methyl ester (C20:0) Behenic acid methyl ester (C22:0)