Weithalsflaschen mit Schraubverschluss, Nalgene®
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2104-0048 2104-0001 2104-0002 2104-0016 2104-0008 2104-0004 2104-0032
215-3981EA 55 EUR InStock
215-3981 215-7201 215-7202 215-7205 215-7204 215-7203 215-7206
Weithalsflaschen mit Schraubverschluss, Nalgene®
Flaschen Kunststoffflaschen
HDPE, translucent with PP screw cap.
  • Good chemical resistance
  • Heat resistant up to 120 °C
  • Leakproof closure
Can be used anywhere in the laboratory and for sample collection. Wide opening allows rapid filling and dispensing of powders, pastes and liquids.
These bottles are non sterile. Ideal for storing, shipping, and packaging liquids, uniform walls provide durability and resistance to splitting or punctures. Manufactured from non-toxic materials.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge