Filter plates, 96-well, His MultiTrap™ TALON®
Lieferant: Cytiva
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
29-0005-96
29-0005-96EA 982 EUR InStock
29-0005-96
Filter plates, 96-well, His MultiTrap™ TALON®
Mikrotiterplatten Mikrofiltrationsplatten
His MultiTrap™ TALON® multiwell plates are designed for high-throughput screening of histidine-tagged proteins from clarified cell lysates. His MultiTrap™ TALON® prepacked plates are manufactured with high reproducibility with regards to the well and media volumes in order to enable consistent results. The average purity of His MultiTrap TALON® is >85% with yields up to 1 mg/well. Leakage of cobalt ions from TALON® Superflow™ medium is negligible. His MultiTrap™ TALON® is used manually with vacuum or centrifugation, or in a liquid handling station.
  • Up to 1 mg of pure histidine-tagged protein per well
  • Large sample volumes can be applied all at once
Prefilled with 50 µl TALON® Superflow™ cobalt-IMAC (immobilized metal affinity chromatography) medium for reproducible and high throughput parallel screening, offering a different selectivity compared to nickel-charged media.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge