Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

1,1'-Diethyl-2,2'-cyanine Iodide 97%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

M-Clarity™ Qualitätsstufe = MQ100

323764-100MG 323764-500MG
323764-100MGEA 78.3 EUR InStock
323764-100MG 323764-500MG
1,1'-Diethyl-2,2'-cyanine Iodide 97%, Sigma-Aldrich®
1,1'-Diethyl-2,2'-cyanine iodide
1,1′-Diethyl-2,2′-cyanine iodide has been used as an absorber in a method to correct the fluorescence excitation-emission matrix (EEM).

1,1′-Diethyl-2,2′-cyanine iodide is a quinocyanine dye. This photographic sensitizing dye is mainly used in silver halide photography. 1,1′-Diethyl-2,2′-cyanine iodide dye aggregation in dilute aqueous solution displays red and blue shifted absorption bands.


Formel: C₂₃H₂₃IN₂
Melting Pt: 273 °C (Zers., Lit.)
Storage Temperature: Room Temperature
MDL Number: MFCD00011971
CAS-Nummer: 977-96-8
EINECS: 213-556-6
ADR: 6.1,II
Order Now
Spezifikation Testergebnisse
Appearance (colour) Dark red to very dark red and dark green to very dark green
Appearance (form) Powder or crystals
Purity (TLC area %) ≥97%
Solubility (colour) Dark red to very dark red
Solubility (turbidity) Clear to very slightly hazy
Solubility (method) 1%, dimethyl formamide:methanol (1:1)
Carbon content 58.6 - 62.9%
Nitrogen content 5.9 - 6.4%
Infrared spectrum Conforms to structure
Wavelength (1) (UV) 520 - 528 nm
Molar absorbancy index (1) ≥40000
Wavelength (2) (UV) 487 - 495 nm
Molar absorbancy index (2) ≥25000
Solvent (UV) Ethanol

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge