Stabilization:
Stabilisiert mit Natriummetaphosphat
33267.09
33267.22
33267.36
33267.09EA
35.1
EUR
InStock
33267.09
33267.22
33267.36
meta-Phosphorsäure 33,5-36,5% stabilisiert ACS
meta-Phosphoric acid
CAS No.: 37267-86-0
Molecular Formula: HPO3
Formula Weight: 79.98
Physical Form: Crystalline
Density: 2.2
Sensitivity: Hygroscopic
Appearance: White
Solubility: Decomposes in water. Soluble in alcohol
Merck Reference: 14,7345
UN No.: 3260
MDL No.: MFCD00011351
Phosphorylating agent, dehydrating agent, dental cements, laboratory reagent