SuperClear® tubes are made of high quality medical grade ultra clear resins to ensure your work is safe and repeatable. These tubes provide a clear view of the tube contents as well as correct colour rendition and fit all standard and high capacity microcentrifuge rotors
- Tubes are available freestanding or with a conical bottom
- ATP, RNase/DNase and endotoxin free (non-pyrogenic)
- Tubes and caps are autoclavable
- Can be centrifuged to 20000 RCF
Available options:
- Tubes with O-ring caps (ethylene propylene)
- Tubes with elastomeric sealing ring
- Tubes only
- Caps only
Verpackung: Packaged in resealable bags. Sterile versions of these tubes are packaged in medical grade bags. Sterilized with a validated medical grade process to an SAL of 10⁻⁶.