39587.02
39587.04
39587.09
39587.02EA
34
EUR
InStock
39587.02
39587.04
39587.09
Vanadium(IV) oxide phthalocyanine
Vanadium(IV) oxide phthalocyanine
CAS No.: 13930-88-6
Molecular Formula: C32H16N8OV
Formula Weight: 579.45
Physical Form: Crystalline
Appearance: Dark blue to purple
UN No.: UN3285
MDL No.: MFCD00041966