39741.03
39741.06
39741.14
39741.03EA
181
EUR
InStock
39741.03
39741.06
39741.14
Gold(I) sodium thiosulphate hydrate ≥99,9% (Metall-Basis)
Gold(I) sodium thiosulphate hydrate
Purity: 99.9%
CAS No.: 15283-45-1
Molecular Formula: Na3Au(S2O3)2.xH2O
Formula Weight: 490.19(anhy)
Physical Form: Powder
Density: 3.09
Appearance: White
Melting Point: dec.
Solubility: Soluble in water. Insoluble in ethanol and most other organic solvents
MDL No.: MFCD00046176