40238.09
40238.18
40238.30
40238.A1
40238.09EA
50.5
EUR
InStock
40238.09
40238.18
40238.30
40238.A1
Strontium molybdenum oxide 99%
Strontium molybdenum oxide
Purity: 99.9%
CAS No.: 13470-04-7
Molecular Formula: SrMoO4
Formula Weight: 247.56
Physical Form: -200 Mesh Powder
Density: 4.540
Melting Point: dec.
Solubility: Insoluble in water. Soluble in acid
MDL No.: MFCD00049550