40458.04
40458.09
40458.04EA
132
EUR
InStock
40458.04
40458.09
Titan(IV)oxid ≥99,9% (Metall-Basis), große Fläche
Titan(IV)oxid
Purity: 99.9%
CAS No.: 13463-67-7
Molecular Formula: O2Ti
Formula Weight: 79.90
Physical Form: -325 Mesh Powder, S.A. 180-300 m2/g
Density: 4.26
Appearance: White
Odour: Odourless
Melting Point: 1830-1850°
Boiling Point: 2500-3000°
Solubility: Insoluble in water, HCl, HNO3, dilute H2SO4; soluble in hot, conc. H2SO4, HF, and alkalies
Merck Reference: 14,9472
MDL No.: MFCD00011269
Photocatalyst, catalyst, catalyst support