41379.04
41379.09
41379.18
41379.04EA
70.4
EUR
InStock
41379.04
41379.09
41379.18
Xylenolorange Tetranatriumsalz ACS
Xylenolorange Tetranatriumsalz
CAS No.: 3618-43-7
Molecular Formula: C31H28N2Na4O13S
Formula Weight: 760.59
Physical Form: Powder
Odour: Odourless
Melting Point: 210° dec.
MDL No.: MFCD00044293