verpackt unter Argon in wiederversiegelbaren ChemSeal™ Flaschen
41842.AK
41842.K2
41842.M1
41842.AKEA
45.5
EUR
InStock
41842.AK
41842.K2
41842.M1
1-Propanol, wasserfrei ≥99,9%
1-Propanol
Purity: 99.9%
CAS No.: 71-23-8
Molecular Formula: CH3CH2CH2OH
Formula Weight: 60.10
Flash Point: 15°(59°F)
Physical Form: Liquid
Density: 0.804
Refractive Index: 1.3850
Odour: Alcohol-like
Melting Point: -127°
Boiling Point: 97°
Beilstein Reference: 1098242
Merck Reference: 14,7842
UN No.: 1274
MDL No.: MFCD00002941