Unter Argon in Ampullen verpackt
43879.06
43879.14
43879.22
43879.06EA
88.9
EUR
InStock
43879.06
43879.14
43879.22
Gallium(III)chlorid, ultra trocken ≥99,999% (Metall-Basis)
Gallium(III)chlorid
Purity: 99.999%
CAS No.: 13450-90-3
Molecular Formula: GaCl3
Formula Weight: 176.03
Physical Form: Crystalline, Ampouled under argon
Density: 2.47
Appearance: White
Melting Point: 78°
Boiling Point: 201°
Merck Reference: 14,4348
UN No.: 3260
MDL No.: MFCD00011018