45577.09
45577.22
45577.36
45577.09EA
26
EUR
InStock
45577.09
45577.22
45577.36
Strontiumperchlorat Trihydrat ≥98%
Strontiumperchlorat Trihydrat
Purity: 98%
CAS No.: 15650-09-6
Molecular Formula: Cl2O8Sr.3H2O
Formula Weight: 340.54 (286.52anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
UN No.: 1508
MDL No.: MFCD09039117