45880.14
45880.22
45880.36
45880.14EA
45.8
EUR
InStock
45880.14
45880.22
45880.36
Zeolite ZSM-5, ammonium
Zeolite ZSM-5, ammonium
CAS No.: 1318-02-1
Molecular Formula: SiO2:Al2O3
Physical Form: Powder, S.A. 400 m2/g, 30:1 mole ratio
Appearance: White
MDL No.: MFCD00132601
Applications: Zeolite ZSM-5 is used in hydrocarbon isomerization, the alkylation of hydrocarbons, and as a catalyst support