Laboratory refrigerator, LABO 100
Lieferant: KIRSCH PHILIPP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
10299
472-0160EA 2820 EUR InStock
472-0160
Laboratory refrigerator, LABO 100
Kühlschränke
This laboratory refrigerator is made of galvanised sheet steel (externally) and impact-resistant, white plastic (internally).
  • Lockable door with easy-to-replace plastic magnetic seal frame
  • Adjustable feet to compensate uneven floors
  • Door hinge on the right-hand side, or the left-hand side, can be retrofitted
  • Condensate evaporates in the refrigerating machine interior
  • Minimum/maximum temperature memory
  • Key switch protected control panel against tampering
  • Alarms can be forwarded using a potential-free contact
  • Anti-freeze against sub-zero temperatures
  • Low-noise compressor reduces noise by 10 to 40 dB
Forced-air cooling with axial fan, switches off automatically when the door opens, ensures a uniform temperature and minimises temperature deviations.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge