Laboratory Ice Crusher
Lieferant: WARING BLENDOR
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
IC20
474-0044EA 1510 EUR InStock
474-0044
Laboratory Ice Crusher
Eiszerkleinerer
The ice crusher is portable, of compact design and especially suitable for countertop use. It has a shaded pole gear motor, 24 rpm. The special height adjustment feature enables the use of any size container up to a maximum of 230 mm high. This is ideal for dispensing ice directly into a blender container.
  • Convenient crushed ice
  • Easily maintained
  • Height adjustment allows direct filling of large containers
Feed chute opening:  38×57 mm
Production of 568 g of ice per minute.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge