Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Test sieves, ASTM E 11, 203×50 mm

Lieferant: Retsch GmbH

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

60.150.000020 60.150.000025 60.150.000032 60.150.000038 60.150.000045 60.150.000053 60.150.000063 60.150.000075 60.150.000090 60.150.000106 60.150.000125 60.150.000150 60.150.000180 60.150.000212 60.150.000250 60.150.000300 60.150.000355 60.150.000425 60.150.000500 60.150.000600 60.150.000710 60.150.000850 60.150.001000 60.150.001180 60.150.001400 60.150.001700 60.150.002000 60.150.002360 60.150.002800 60.150.003350 60.150.004000 60.150.004750 60.150.005600 60.150.006300 60.150.006700 60.150.008000 60.150.009500 60.150.011200 60.150.012500 60.150.013200 60.150.016000 60.150.019000 60.150.022400 60.150.025000 60.150.026500 60.150.031500 60.150.037500 60.150.045000 60.150.050000 60.150.053000 60.150.063000 60.150.075000 60.150.090000 60.150.100000 60.150.106000 60.150.125000
510-5321EA 838 EUR InStock
510-5321 510-5322 510-5323 510-5324 510-5325 510-5326 510-5327 510-5328 510-5329 510-5330 510-5331 510-5332 510-5333 510-5334 510-5335 510-5336 510-5337 510-5338 510-5339 510-5340 510-5341 510-5342 510-5343 510-5344 510-5345 510-5346 510-5347 510-5348 510-5349 510-5350 510-5351 510-5352 510-5353 510-5354 510-5355 510-5356 510-5357 510-5358 510-5359 510-5360 510-5361 510-5362 510-5363 510-5364 510-5365 510-5366 510-5367 510-5368 510-5369 510-5370 510-5371 510-5372 510-5373 510-5374 510-5375 510-5376
Test sieves, ASTM E 11, 203×50 mm
Siebe Analysensiebe
Sieves made from 304 stainless steel are individually laser engraved with clear and precise labelling for full traceability. They are fully compatible with Retsch's existing products and can be combined with other sieves without problems.

  • Sieve bodies are made in one piece and the absence of hollow spaces prevents cross-contamination
  • High corrosion resistance and easy to clean
  • Maximum stability and optimal sealing of the sieve stack, 15% less weight than traditional sieves

Sieve meshes, frames and labeling comply with standards. The sieves are tested 5 times and are delivered with test report.

Zertifizierungen: An individual inspection certificate for test materials monitoring according to ASTM E 11 is also available on request.

Bestellinformation: Sieves and accessories of other dimensions are available on request. Also available with perforated plate, round or square. For a full overview, please go to website.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge